1-Ethyl-3-methylimidazolium-p-toluenesulphonate ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 1-Ethyl-3-methylimidazolium tosylate, 1-Ethyl-3-methyl-1H-imidazol-3-ium 4-methylbenzenesulphonate, [EMIM] [TOS]

E0651-5G E0651-25G
TCIAE0651-5GEA 905 NOK
TCIAE0651-5G TCIAE0651-25G
1-Ethyl-3-methylimidazolium-p-toluenesulphonate ≥98.0% (by HPLC)
1-Ethyl-3-methylimidazolium-p-toluenesulphonate
Formel: C₁₃H₁₈N₂O₃S
MW: 282,36 g/mol
MDL nummer: MFCD03788929
CAS-nummer: 328090-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55b9e9526d45db2c956e7f10b1253e3e3c02613d&vid=55b9e9526d45db2c956e7f10b1253e3e3c02613d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Ion exchange titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR