3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2,3-Dihydrothieno[3,4-b]-1,4-dioxine-5,7-dicarboxylic Acid, 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5,7-dicarboxylic acid

E0743-1G E0743-5G
TCIAE0743-1GEA 730 NOK
TCIAE0743-1G TCIAE0743-5G
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid ≥95.0% (by GC, titration analysis)
3,4-Ethylenedioxythiophene-2,5-dicarboxylic acid
Formel: C₈H₆O₆S
MW: 230,19 g/mol
MDL nummer: MFCD00966477
CAS-nummer: 18361-03-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR