2-Ethoxy-5-(1-propenyl)phenol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Ethoxy-2-hydroxy-4-(1-propenyl)benzene, Propenylguaethol

E0804-25G E0804-100G
TCIAE0804-25GEA 620 NOK
TCIAE0804-25G TCIAE0804-100G
2-Ethoxy-5-(1-propenyl)phenol ≥98.0% (by GC)
2-Ethoxy-5-(1-propenyl)phenol
Formel: C₁₁H₁₄O₂
MW: 178,23 g/mol
Kokepunkt: 116°C
MDL nummer: MFCD00009283
CAS-nummer: 94-86-0
EINECS: 202-370-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR