Ethyl-2-bromothiazole-5-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Ethyl 2-bromothiazole-5-carboxylate
E0891-1G
E0891-5G
TCIAE0891-1GEA
570
NOK
TCIAE0891-1G
TCIAE0891-5G
Ethyl-2-bromothiazole-5-carboxylate ≥98.0% (by GC)
Ethyl-2-bromothiazole-5-carboxylate
Formel:
C₆H₆BrNO₂S MW: 236,08 g/mol Kokepunkt: 68°C |
MDL nummer:
MFCD00463837 CAS-nummer: 41731-83-3 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...