2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 4-Carboxymethyl-2-ethoxybenzoic Acid Ethyl Ester, Ethyl 4-Carboxymethyl-2-ethoxybenzoate, 2-(3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic acid

E0928-5G E0928-25G
TCIAE0928-5GEA 945 NOK
TCIAE0928-5G TCIAE0928-25G
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid ≥98.0% (by GC, titration analysis)
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid
Formel: C₁₃H₁₆O₅
MW: 252,27 g/mol
MDL nummer: MFCD08704233
CAS-nummer: 99469-99-5
EINECS: 427-630-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR