Emedastine difumarate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(2-Ethoxyethyl)-2-(4-methyl-1,4-diazepan-1-yl)-1H-benzo[d]imidazole difumarate, 1-(2-Ethoxyethyl)-2-(4-methyl-1-homopiperazinyl)benzimidazole Difumarate
E0936-200MG
E0936-1G
TCIAE0936-200MGEA
1875
NOK
TCIAE0936-200MG
TCIAE0936-1G
Emedastine difumarate ≥98.0% (by HPLC, titration analysis)
Emedastine difumarate
Formel:
C₁₇H₂₆N₄O MW: 534,57 g/mol |
MDL nummer:
MFCD00923844 CAS-nummer: 87233-62-3 Merck Indeks: 14,03557 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...