Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester, Ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate

E0980-5G E0980-25G
TCIAE0980-5GEA 220 NOK
TCIAE0980-5G TCIAE0980-25G
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate ≥98.0% (by GC)
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate
Formel: C₁₂H₁₄N₂O₄S
MW: 282,31 g/mol
MDL nummer: MFCD00832845
CAS-nummer: 58168-20-0

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 137.0 to 141.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR