Ethyl (4-methylphenoxy)acetate ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(4-Methylphenoxy)acetic Acid Ethyl Ester, Ethyl 2-(p-tolyloxy)acetate
E1015-5G
E1015-25G
TCIAE1015-5GEA
405
NOK
TCIAE1015-5G
TCIAE1015-25G
Ethyl (4-methylphenoxy)acetate ≥97.0% (by GC)
Ethyl (4-methylphenoxy)acetate
Formel:
C₁₁H₁₄O₃ MW: 194,23 g/mol Kokepunkt: 152°C |
MDL nummer:
MFCD00209535 CAS-nummer: 67028-40-4 EINECS: 266-554-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...