Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
Ethyl 3-(3-amino-4-(methylamino)-N-(pyridin-2-yl)benzamido)propanoate, 3-[3-Amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionic Acid Ethyl Ester
E1033-1G
E1033-5G
TCIAE1033-1GEA
245
NOK
TCIAE1033-1G
TCIAE1033-5G
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate ≥98.0% (by HPLC, titration analysis)
Ethyl-3-[3-amino-4-(methylamino)-N-(2-pyridyl)benzamido]propionate
Formel:
C₁₈H₂₂N₄O₃ MW: 342,40 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD09833624 CAS-nummer: 212322-56-0 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...