1-Ethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 1-Ethylpyrazole
E1037-5ML E1037-25ML
TCIAE1037-5MLEA 350 NOK
TCIAE1037-5ML TCIAE1037-25ML
1-Ethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-Ethyl-1H-pyrazole
Formel: C₅H₈N₂
MW: 96,1304 g/mol
Kokepunkt: 137°C
MDL nummer: MFCD03444367
CAS-nummer: 2817-71-2
FN: 1993
ADR: 3,III
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %