Ethyl 3-Oxo-3-(2-thienyl)propionate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Ethyl 3-oxo-3-(thiophen-2-yl)propanoate, 3-Oxo-3-(2-thienyl)propionic acid Ethyl ester

E1073-200MG E1073-1G
TCIAE1073-200MGEA 325 NOK
TCIAE1073-200MG TCIAE1073-1G
Ethyl 3-Oxo-3-(2-thienyl)propionate ≥98.0% (by GC)
Ethyl 3-Oxo-3-(2-thienyl)propionate
Formel: C₉H₁₀O₃S
MW: 198,24 g/mol
Kokepunkt: 130°C
MDL nummer: MFCD00542649
CAS-nummer: 13669-10-8

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR