5-Ethynyl-2'-deoxycytidine 98,0 N_ASSAY_METHOD

Leverandør: TCI

Synonymer: 4-Amino-5-ethynyl-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 5'-Ethynyl-2'-deoxycytidine, EdC

E1093-50MG E1093-200MG
TCIAE1093-50MGEA 1675 NOK
TCIAE1093-50MG TCIAE1093-200MG
5-Ethynyl-2'-deoxycytidine 98,0 N_ASSAY_METHOD
5-Ethynyl-2'-deoxycytidine
Formel: C₁₁H₁₃N₃O₄
MW: 251.24 g/mol
MDL nummer: MFCD22417249
CAS-nummer: 69075-47-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR