Ethyl 4-Fluorocinnamate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4-Fluorocinnamic acid ethyl ester, Ethyl (2E)-3-(4-fluorophenyl)acrylate

E1099-1G E1099-5G
TCIAE1099-1GEA 1775 NOK
TCIAE1099-1G TCIAE1099-5G
Ethyl 4-Fluorocinnamate ≥98.0% (by GC)
Ethyl 4-Fluorocinnamate
Formel: C₁₁H₁₁FO₂
MW: 194,21 g/mol
Kokepunkt: 131°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02252626
CAS-nummer: 352-03-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3cf8fa31801a9f720f7d5e7603f13d34a945e9a&vid=e667e261b6136f585187ba4a6e62b1d12abe3fff&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR