Ethyl (4-bromobenzoyl)acetate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
ethyl 3-(4-bromophenyl)-3-oxopropanoate, 3-(4-bromophenyl)-3-oxo-propionic acid ethyl ester, Benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester
E1117-1G
E1117-5G
TCIAE1117-1GEA
1350
NOK
TCIAE1117-1G
TCIAE1117-5G
Ethyl (4-bromobenzoyl)acetate ≥98.0% (by GC)
Ethyl (4-bromobenzoyl)acetate
Formel:
C₁₁H₁₁BrO₃ MW: 271,11 g/mol Kokepunkt: 124°C |
MDL nummer:
MFCD00231243 CAS-nummer: 26510-95-2 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...