Ethyl-5-amino-1-benzofuran-2-carboxylate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
Ethyl 5-Aminobenzofuran-2-carboxylate, Ethyl 5-aminobenzo[b]furan-2-carboxylate, 5-Aminobenzofuran-2-carboxylic acid ethyl ester
E1138-1G
E1138-5G
TCIAE1138-1GEA
405
NOK
TCIAE1138-1G
TCIAE1138-5G
Ethyl-5-amino-1-benzofuran-2-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-5-amino-1-benzofuran-2-carboxylate
Formel:
C₁₁H₁₁NO₃ MW: 205,22 g/mol |
MDL nummer:
MFCD08275091 CAS-nummer: 174775-48-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...