Ethyl-2-acetyl-3-ethoxyacrylate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Ethyl 2-(ethoxymethylene)acetoacetate, Ethyl 2-(ethoxymethylene)-3-oxobutanoate

E1148-5G E1148-25G
TCIAE1148-5GEA 460 NOK
TCIAE1148-5G TCIAE1148-25G
Ethyl-2-acetyl-3-ethoxyacrylate ≥98.0% (by GC)
Ethyl-2-acetyl-3-ethoxyacrylate
Formel: C₉H₁₄O₄
MW: 186,21 g/mol
Kokepunkt: 266°C
MDL nummer: MFCD09959866
CAS-nummer: 3788-94-1
EINECS: 223-259-3

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR