(9-Ethyl-9H-carbazol-3-yl)boronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 9-Ethylcarbazole-3-boronic acid

E1153-1G E1153-5G
TCIAE1153-1GEA 555 NOK
TCIAE1153-1G TCIAE1153-5G
(9-Ethyl-9H-carbazol-3-yl)boronic acid (contains varying amounts of Anhydride)
(9-Ethyl-9H-carbazol-3-yl)boronic acid
Formel: C₁₄H₁₄BNO₂
MDL nummer: MFCD09743121
CAS-nummer: 669072-93-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR