Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
Ethyl 4-(4-Methylphenyl)-4-oxobutanoate, 4-Oxo-4-(p-tolyl)butyric acid ethyl ester, Ethyl 4-oxo-4-(p-tolyl)butanoate, 4-(4-Methylphenyl)-4-oxobutanoic acid ethyl ester, Ethyl 4-Oxo-4-(p-tolyl)butyrate
E1195-1G
E1195-5G
TCIAE1195-1GEA
1625
NOK
TCIAE1195-1G
TCIAE1195-5G
Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95.0% (by GC)
Ethyl-4-(4-methylphenyl)-4-oxobutyrate
Formel:
C₁₃H₁₆O₃ MW: 220,27 g/mol Kokepunkt: 178°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00266564 CAS-nummer: 6942-61-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 95.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...