Ethyl (2-Methoxyphenoxy)acetate ≥98.0% (by GC)
Leverandør: TCI
Synonymer: (2-Methoxyphenoxy)acetic acid Ethyl ester, Ethyl 2-(2-methoxyphenoxy)acetate
E1241-5G E1241-25G
TCIAE1241-5GEA 835 NOK
TCIAE1241-5G TCIAE1241-25G
Ethyl (2-Methoxyphenoxy)acetate ≥98.0% (by GC)
Ethyl (2-Methoxyphenoxy)acetate
Formel: C₁₁H₁₄O₄
MW: 210,23 g/mol
Kokepunkt: 116°C
MDL nummer: MFCD00082657
CAS-nummer: 13078-21-2
EINECS: 235-977-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f478529d25a9ac0129386901c3af14b2c661ede&vid=7f478529d25a9ac0129386901c3af14b2c661ede&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %