Ethyl azidoacetate ≥95.0% (by GC)
Leverandør: TCI
Warning
Synonymer: (2-ethoxy-2-oxoethyl)imino-iminoammonium, Ethyl 2-(1H-triazirin-1-yl)acetate, Acetic acid, 2-azido-, ethyl ester
E1255-5G E1255-25G
TCIAE1255-5GEA 1025 NOK
TCIAE1255-5G TCIAE1255-25G
Ethyl azidoacetate ≥95.0% (by GC)
Ethyl azidoacetate
Formel: C₄H₇N₃O₂
MW: 129,12 g/mol
Kokepunkt: 164°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00190177
CAS-nummer: 637-81-0
EINECS: 211-301-3
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89cb93c19f163349894990844ad0e30a9abb158a&vid=89cb93c19f163349894990844ad0e30a9abb158a&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %