Ethyl 2-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]acetate ≥98.0% (by HPLC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]acetic acid Ethyl ester
E1268-100MG
E1268-500MG
TCIAE1268-100MGEA
1300
NOK
TCIAE1268-100MG
TCIAE1268-500MG
Ethyl 2-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]acetate ≥98.0% (by HPLC)
Ethyl 2-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]acetate
Formel:
C₂₅H₃₀NO₆P MW: 471,49 g/mol Lagringstemperatur: Fryseskap |
CAS-nummer:
2173111-94-7 |
Spesifikasjon Testresultater
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 2.70 to 3.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...