Ethyl 6-Hydroxyhexanoate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: 6-Hydroxyhexanoic acid Ethyl ester, Ethyl 6-hydroxyhexanoate

E1305-5G E1305-25G
TCIAE1305-5GEA 1225 NOK
TCIAE1305-5G TCIAE1305-25G
Ethyl 6-Hydroxyhexanoate ≥95.0% (by GC)
Ethyl 6-Hydroxyhexanoate
Formel: C₈H₁₆O₃
MW: 160,21 g/mol
Kokepunkt: 128°C
MDL nummer: MFCD00075601
CAS-nummer: 5299-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ab46ba0bca8fbd3fc7993a06f8df26388c21d9a&vid=8ab46ba0bca8fbd3fc7993a06f8df26388c21d9a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR