Ethyl trimethylacetopyruvate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Ethyl 5,5-dimethyl-2,4-dioxohexanoate

E1326-1G E1326-5G
TCIAE1326-1GEA 700 NOK
TCIAE1326-1G TCIAE1326-5G
Ethyl trimethylacetopyruvate ≥98.0% (by GC)
Ethyl trimethylacetopyruvate
Formel: C₁₀H₁₆O₄
MW: 200,24 g/mol
Kokepunkt: 124°C
MDL nummer: MFCD00052319
CAS-nummer: 13395-36-3
EINECS: 236-478-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR