2-Ethyl-1H-benzo[d]imidazole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
2-Ethyl-1H-benzimidazole
E1358-5G
E1358-25G
TCIAE1358-5GEA
755
NOK
TCIAE1358-5G
TCIAE1358-25G
2-Ethyl-1H-benzo[d]imidazole ≥98.0% (by GC, titration analysis)
2-Ethyl-1H-benzo[d]imidazole
Formel:
C₉H₁₀N₂ MW: 146,1900 g/mol |
MDL nummer:
MFCD00022684 CAS-nummer: 1848-84-6 EINECS: 217-433-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 173.0 to 177.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...