Etofibrate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl Nicotinate, Nicotinic acid 2-[[2-(4-Chlorophenoxy)-2-methylpropanoyl]oxy]ethyl ester, 2-((2-(4-Chlorophenoxy)-2-methylpropanoyl)oxy)ethyl nicotinate, 3-Pyridinecarboxylic acid 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl ester, 2-[2-(4-Chlorophenoxy)-2-methyl-1-oxopropoxy]ethyl 3-Pyridinecarboxylate

E1359-50MG E1359-250MG
TCIAE1359-50MGEA 1400 NOK
TCIAE1359-50MG TCIAE1359-250MG
Etofibrate ≥98.0% (by GC)
Etofibrate
Formel: C₁₈H₁₈ClNO₅
MW: 363,79 g/mol
Kokepunkt: 214°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00865727
CAS-nummer: 31637-97-5
EINECS: 250-743-1
Merck Indeks: 14,03877

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR