EPI-001 ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Bisphenol A (3-Chloro-2-hydroxypropyl)(2,3-dihydroxypropyl)ether, 3-[4-[2-[4-(3-Chloro-2-hydroxypropoxy)phenyl]propan-2-yl]phenoxy]propane-1,2-diol, 3-(4-(2-(4-(3-Chloro-2-hydroxypropoxy)phenyl)propan-2-yl)phenoxy)propane-1,2-diol

E1380-5MG E1380-25MG
TCIAE1380-5MGEA 890 NOK
TCIAE1380-5MG TCIAE1380-25MG
EPI-001 ≥98.0% (by GC)
EPI-001
Formel: C₂₁H₂₇ClO₅
MW: 394,89 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02683414
CAS-nummer: 227947-06-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR