Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: Ethyl 2-((ethoxycarbonothioyl)thio)propanoate, 2-[(Ethoxycarbonothioyl)thio]propionic acid Ethyl ester, Ethyl 2-[(ethoxycarbonothioyl)thio]propanoate, 2-[(Ethoxycarbonothioyl)thio]propanoic acid Ethyl ester

E1403-1G E1403-5G
TCIAE1403-1GEA 2525 NOK
TCIAE1403-1G TCIAE1403-5G
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate ≥95.0% (by GC)
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate
Formel: C₈H₁₄O₃S₂
MW: 222,32 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD30474932
CAS-nummer: 73232-07-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.1240 to 1.1340

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR