Ethyl acetoacetate propylene glycol ketal ≥98.0% (by GC)
Leverandør: TCI
Synonymer: Ethyl Acetoacetate Propylene glycol Ketal, 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetic acid ethyl ester, Ethyl 2-(2,4-Dimethyl-1,3-dioxolan-2-yl)acetate (mixture of diastereoisomers)
E1417-100G E1417-25G
TCIAE1417-100GEA 785 NOK
TCIAE1417-100G TCIAE1417-25G
Ethyl acetoacetate propylene glycol ketal ≥98.0% (by GC)
Ethyl acetoacetate propylene glycol ketal
Formel: C₉H₁₆O₄
MW: 188,23 g/mol
Kokepunkt: 76°C
Flash punkt: 65°C
MDL nummer: MFCD00151819
CAS-nummer: 6290-17-1
EINECS: 228-536-2
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 % (total of isomer)