N-Formyl-L-tyrosine ≥98.0% (by titrimetric analysis)

Leverandør: TCI

F0135-1G F0135-5G
TCIAF0135-1GEA 1225 NOK
TCIAF0135-1G TCIAF0135-5G
N-Formyl-L-tyrosine ≥98.0% (by titrimetric analysis)
N-Formyl-L-tyrosine
Formel: C₁₀H₁₁NO₄
MW: 209,20 g/mol
MDL nummer: MFCD00021031
CAS-nummer: 13200-86-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +81.0 to +85.0 deg (C=2.6, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR