N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

F0358-25G F0358-5G F0358-100G
TCIAF0358-25GEA 1175 NOK
TCIAF0358-25G TCIAF0358-5G TCIAF0358-100G
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
Formel: C₇H₁₄B₂ClF₉N₂
MW: 354,26 g/mol
MDL nummer: MFCD00142607
CAS-nummer: 140681-55-6
EINECS: 414-380-4
FN: 3088
ADR: 4.2,II
Merck Indeks: 14,08425

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR