(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Fmoc-Glu-OH, N-Fmoc-L-glutamic Acid

F0453-1G F0453-5G
TCIAF0453-1GEA 220 NOK
TCIAF0453-1G TCIAF0453-5G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid
Formel: C₂₀H₁₉NO₆
MW: 369,4 g/mol
MDL nummer: MFCD00237657
CAS-nummer: 121343-82-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -25.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR