1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: PPZ

F0650-1G F0650-100MG
TCIAF0650-1GEA 6350 NOK
TCIAF0650-1G TCIAF0650-100MG
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine
Formel: C₁₁H₁₃FN₄O₄
MW: 284,25 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 928830-73-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Amber powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR