4-Fluoro-2,1,3-benzoxadiazole ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4-Fluorobenzofurazan

F0702-1G F0702-5G
TCIAF0702-1GEA 1475 NOK
TCIAF0702-1G TCIAF0702-5G
4-Fluoro-2,1,3-benzoxadiazole ≥98.0% (by GC)
4-Fluoro-2,1,3-benzoxadiazole
Formel: C₆H₃FN₂O
MW: 138,10 g/mol
Kokepunkt: 83°C
MDL nummer: MFCD03425566
CAS-nummer: 29270-55-1

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR