Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥97.0% (by total nitrogen basis)

Leverandør: TCI
Warning

Synonymer: TFFH, N-((Dimethylamino)fluoromethylene)-N-methylmethanaminium hexafluorophosphate(V)

F0726-1G F0726-5G
TCIAF0726-1GEA 635 NOK
TCIAF0726-1G TCIAF0726-5G
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥97.0% (by total nitrogen basis)
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
Formel: C₅H₁₂F₇N₂P
MW: 264,13 g/mol
MDL nummer: MFCD02684443
CAS-nummer: 164298-23-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR