N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Nα-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH, N-Fmoc-S-trityl-D-cysteine

F0752-5G F0752-1G
TCIAF0752-5GEA 875 NOK
TCIAF0752-5G TCIAF0752-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine
Formel: C₃₇H₃₁NO₄S
MW: 585,72 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00151922
CAS-nummer: 167015-11-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Specific rotation [a]20/D -15.0 to -19.0 deg (C=1, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR