(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Nalpha-Fmoc-O-tert-butyl-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, N-Fmoc-O-tert-butyl-D-tyrosine

F0772-1G F0772-5G
TCIAF0772-1GEA 435 NOK
TCIAF0772-1G TCIAF0772-5G
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥98.0% (by HPLC, titration analysis)
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
Formel: C₂₈H₂₉NO₅
MW: 459,5 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00065684
CAS-nummer: 118488-18-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C
Specific rotation [a]20/D +28.0 to +32.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR