3'-Fluoro-4'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

F0843-1G F0843-5G
TCIAF0843-1GEA 795 NOK
TCIAF0843-1G TCIAF0843-5G
3'-Fluoro-4'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)
3'-Fluoro-4'-hydroxyacetophenone
Formel: C₈H₇FO₂
MW: 154,14 g/mol
MDL nummer: MFCD00215840
CAS-nummer: 403-14-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 125.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR