2-Fluor-5-trifluormetylpyridin ≥98.0% (by GC)

Leverandør: TCI
Warning

F0846-1G F0846-5G
TCIAF0846-1GEA 230 NOK
TCIAF0846-1G TCIAF0846-5G
2-Fluor-5-trifluormetylpyridin ≥98.0% (by GC)
2-Fluor-5-trifluormetylpyridin
Formel: C₆H₃F₄N
MW: 165,09 g/mol
Kokepunkt: 120°C
Flash punkt: 32°C
MDL nummer: MFCD07437945
CAS-nummer: 69045-82-5
EINECS: 400-290-2
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR