3-Fluoro-4-morpholinoaniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-(2-Fluoro-4-aminophenyl)morpholine

F0849-5G F0849-25G
TCIAF0849-5GEA 1050 NOK
TCIAF0849-5G TCIAF0849-25G
3-Fluoro-4-morpholinoaniline ≥98.0% (by GC, titration analysis)
3-Fluoro-4-morpholinoaniline
Formel: C10H13FN2O
MW: 196,2214 g/mol
MDL nummer: MFCD02571270
CAS-nummer: 93246-53-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR