4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulphonylamino)-5-pyrimidinecarboxaldehyde ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulphonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulphonamide

F0867-5G F0867-25G
TCIAF0867-5GEA 350 NOK
TCIAF0867-5G TCIAF0867-25G
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulphonylamino)-5-pyrimidinecarboxaldehyde ≥98.0% (by HPLC, total nitrogen)
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulphonylamino)-5-pyrimidinecarboxaldehyde
Formel: C₁₆H₁₈FN₃O₃S
MW: 351,40 g/mol
MDL nummer: MFCD08458342
CAS-nummer: 147118-37-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR