(9H-Fluoren-9-yl)methyl hydrazinecarboxylate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Fmoc-hydrazine, 9-Fluorenylmethyl Carbazate, [(9H-Fluoren-9-ylmethoxy)carbonyl]hydrazine, Carbazic acid 9-Fluorenylmethyl ester

F0872-5G F0872-25G
TCIAF0872-5GEA 380 NOK
TCIAF0872-5G TCIAF0872-25G
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate ≥98.0% (by HPLC, titration analysis)
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate
Formel: C₁₅H₁₄N₂O₂
MW: 254,2800 g/mol
Smeltepunkt: 171°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00239419
CAS-nummer: 35661-51-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR