2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
|
Warning
|
Synonymer:
Fmoc-Glycinol, N-Fmoc-ethanolamine, 2-(Fmoc-amino)-1-ethanol
F0912-5G
F0912-25G
TCIAF0912-5GEA
380
NOK
TCIAF0912-5G
TCIAF0912-25G
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol ≥98.0% (by HPLC, total nitrogen)
2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol
Formel:
C₁₇H₁₇NO₃ MW: 283,33 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00235927 CAS-nummer: 105496-31-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...