2-Fluoro-6-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 2-Fluoro-6-methoxybenzeneboronic acid

F0941-1G F0941-5G
TCIAF0941-1GEA 245 NOK
TCIAF0941-1G TCIAF0941-5G
2-Fluoro-6-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-6-methoxyphenylboronic acid
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL nummer: MFCD02179483
CAS-nummer: 78495-63-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR