3-Fluoro-4-methoxybenzyl chloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 3-Fluoro-4-methoxybenzylchloride

F0966-5G F0966-25G
TCIAF0966-5GEA 795 NOK
TCIAF0966-5G TCIAF0966-25G
3-Fluoro-4-methoxybenzyl chloride ≥98.0% (by GC, titration analysis)
3-Fluoro-4-methoxybenzyl chloride
Formel: C₈H₈ClFO
MW: 174,60 g/mol
Kokepunkt: 62°C
MDL nummer: MFCD04973773
CAS-nummer: 351-52-0
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR