Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI

F0973-1G F0973-5G F0973-25G
TCIAF0973-1GEA 635 NOK
TCIAF0973-1G TCIAF0973-5G TCIAF0973-25G
Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)
Formamidine hydrobromide
Formel: CH₄N₂·HBr
MDL nummer: MFCD28369274
CAS-nummer: 146958-06-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR