3-Fluoro-4-formylphenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Fluoro-4-formylbenzeneboronic acid
F1051-1G
F1051-5G
TCIAF1051-1GEA
525
NOK
TCIAF1051-1G
TCIAF1051-5G
3-Fluoro-4-formylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-4-formylphenylboronic acid
Formel:
C₇H₆BFO₃ MW: 167,93 g/mol |
MDL nummer:
MFCD02093051 CAS-nummer: 248270-25-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...