2-Fluoroethyl p-Toluenesulphonate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Fluoroethyl Tosylate, 2-Fluoroethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-Fluoroethyl ester

F1056-1G F1056-5G
TCIAF1056-1GEA 890 NOK
TCIAF1056-1G TCIAF1056-5G
2-Fluoroethyl p-Toluenesulphonate ≥98.0% (by GC)
2-Fluoroethyl p-Toluenesulphonate
Formel: C₉H₁₁FO₃S
MW: 218,24 g/mol
Kokepunkt: 140°C
MDL nummer: MFCD01658061
CAS-nummer: 383-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd3bf857c20f920f55cf7a04399cfc71b3e5f825&vid=cd3bf857c20f920f55cf7a04399cfc71b3e5f825&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR