4-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Fluoro-2-methoxybenzeneboronic acid
F1062-1G
F1062-5G
TCIAF1062-1GEA
650
NOK
TCIAF1062-1G
TCIAF1062-5G
4-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-methoxyphenylboronic acid
Formel:
C₇H₈BFO₃ MW: 169,95 g/mol |
MDL nummer:
MFCD03788423 CAS-nummer: 179899-07-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...