3-Fluoro-5-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 3-Fluoro-5-methoxybenzeneboronic acid

F1063-1G F1063-5G
TCIAF1063-1GEA 1075 NOK
TCIAF1063-1G TCIAF1063-5G
3-Fluoro-5-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-5-methoxyphenylboronic acid
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL nummer: MFCD07363780
CAS-nummer: 609807-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5445aaf83bd3d45d6395b199baf482eb57036f37&vid=5445aaf83bd3d45d6395b199baf482eb57036f37&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR