2-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid

F1071-1G F1071-5G
TCIAF1071-1GEA 745 NOK
TCIAF1071-1G TCIAF1071-5G
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formel: C₈H₈BFO₄
MW: 197,96 g/mol
Smeltepunkt: 140°C
MDL nummer: MFCD08436034
CAS-nummer: 603122-84-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR